Learn More
Nitinol compression coil spring, wire diameter 1.0mm (0.039in), mean coil diameter 7.0mm (0.27in), Alloy M, Thermo Scientific™
£63.70 - £881.00
Specifications
Chemical Name or Material | Nitinol compression coil spring, Alloy M |
---|---|
Name Note | Wire diameter 1.0mm (0.039 in.), mean coil diameter 7.0mm (0.27 in.) |
MDL Number | MFCD02091734 |
Odor | Odorless |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15473995
|
Thermo Scientific Alfa Aesar
045578.KJ |
5 Pc. | |||||||
15483995
|
Thermo Scientific Alfa Aesar
045578.KM |
25 Pc. |
£272.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15493995
|
Thermo Scientific Alfa Aesar
045578.KN |
100 Pc. | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Nitinol compression coil spring, Alloy M | |
Wire diameter 1.0mm (0.039 in.), mean coil diameter 7.0mm (0.27 in.) | |
MFCD02091734 | |
Odorless |
Safety and Handling
GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures