Learn More
Nitinol foil, 0.127mm (0.005in) thick, shape memory, flat annealed, pickled surface, Thermo Scientific Chemicals
£88.50 - £914.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15423995
|
Thermo Scientific Alfa Aesar
045514.FF |
25 x 25 mm |
£88.50
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15433995
|
Thermo Scientific Alfa Aesar
045514.FI |
50 x 50 mm |
£281.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15443995
|
Thermo Scientific Alfa Aesar
045514.GH |
100 x 100 mm |
£914.00
Each |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Nitinol foil is used in cell phone antennas, children's toys, experimental materials, eye glasses components and medical equipment. It is also used in heat engines, resilient glass frames, mechanical watch springs, temperature control system, couplings, golf club insert, biomedical and medical uses, cell-phone technology: retractable antenna, microphone boom. It is highly biocompatible and has properties suitable for use in orthopaedic implants.