Learn More
Ansell™ NitraSafe™ 28-329 Series Nitrile Heavy Weight Gloves
Product Code. 12323589
Click to view available options
Size:
8
9
10
Combines the outstanding protection of Kevlar™-reinforced construction with the grip and flexibility of Ansell’s advanced nitrile coating technology. The NitraSafe™ range stands for safe handling of oily heavy metal parts, while the NitraSafe™ Foam combines good grip and comfort.
  • Provides safe handling of oily heavy metal parts, while the NitraSafe™ Foam combines good grip and comfort.
  • Both gloves, possibly in combination with the NitraGold™ cuff, offer outstanding mechanical protection against cuts and abrasion.
  • Cut and sewn construction
  • Cotton Jersey Kevlar™ construction
  • Nitrile coating
  • Palm-coated finish
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Certifications/Compliance CE Mark 0493, Standard Category III EN 388 : 4331
Cuff Material Nitrile
Product Type Heavyweight Gloves
Size 9
Glove Cuff Style Nitragold™
Liner Material Cotton Jersey Dupont™ Kevlar™ aramid fiber
Lining Cotton Jersey Dupont™ Kevlar™ aramid fiber
Palm Type Coated-palm
Quantity 72 Pairs
Resistant To Greater resistance to cuts and abrasion