Learn More
Ansell™ NitraSafe™ 28-329 Series Nitrile Heavy Weight Gloves
Combines the outstanding protection of Kevlar™-reinforced construction with the grip and flexibility of Ansell’s advanced nitrile coating technology. The NitraSafe™ range stands for safe handling of oily heavy metal parts, while the NitraSafe™ Foam combines good grip and comfort.
£221.00
Specifications
Back Material Nitrile
Certifications/Compliance CE Mark 0493, Standard Category III EN 388 : 4331
Cuff Material Nitrile
Product Type Heavyweight Gloves
Glove Cuff Style Nitragold™
View More Specs
Products 3
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12313589
View Documents
Ansell™
28329080
8
£221.00
12 pairs
Estimated Shipment: 23-07-2025
to see stock.
Add to basket
12323589
View Documents
Ansell™
28329090
9
£221.00
12 pairs
Estimated Shipment: 23-07-2025
to see stock.
Add to basket
12600812
View Documents
Ansell™
28329100
10
£221.00
12 pairs
Estimated Shipment: 23-07-2025
to see stock.
Add to basket
Description
Description
  • Provides safe handling of oily heavy metal parts, while the NitraSafe™ Foam combines good grip and comfort.
  • Both gloves, possibly in combination with the NitraGold™ cuff, offer outstanding mechanical protection against cuts and abrasion.
  • Cut and sewn construction
  • Cotton Jersey Kevlar™ construction
  • Nitrile coating
  • Palm-coated finish
Specifications
Specifications
Nitrile
Nitrile
Nitragold™
Cotton Jersey Dupont™ Kevlar™ aramid fiber
72 Pairs
CE Mark 0493, Standard Category III EN 388 : 4331
Heavyweight Gloves
Cotton Jersey Dupont™ Kevlar™ aramid fiber
Coated-palm
Greater resistance to cuts and abrasion
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Ansell™ NitraSafe™ 28-329 Series Nitrile Heavy Weight Gloves