Learn More
Ansell™ NitraSafe™ 28-360 Series Nitrile Heavy Weight Gloves
Product Code. 12690812
Click to view available options
Size:
8
9
10
Brown Nitrile Heavy Weight Gloves combine the outstanding protection of Kevlar™-reinforced construction with the grip and flexibility of Ansell’s advanced nitrile coating technology.
  • Industrial gloves
  • NitraSafe™ range stands for safe handling of oily heavy metal parts
  • Mechanical protection against cuts and abrasion
  • Heavy weight design provides feeling of cushioning for the wearer
  • Protective gloves combines the protection of Kevlar™ reinforced construction and grip, flexibility of advanced nitrile coating technology
Applications:
  • Handling parts with sharp, rough edges
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile Foam
Certifications/Compliance CE Mark 0493, Standard Category III EN 388 : 3321
Cuff Material Nitrile
Product Type Heavyweight Gloves
Size 8
Glove Cuff Style Nitragold™
Liner Material Cotton Jersey Dupont™ Kevlar™ aramid fiber
Lining Cotton Jersey Dupont™ Kevlar™ aramid fiber
Palm Type Fully coated-palm
Quantity 72 Pairs
Resistant To Greater resistance to cuts and abrasion