missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrate Anion Standard, 0.2mg/ml, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
This standard is prepared, tested, certified and verified by following the exact same regime as already presented for ICP-MS Standards. It delivers confidence in the reliability of ion chromatography analysis. All are prepared from High Purity Starting materials and are traceable to N.I.S.T, where possible.
Specifications
Specifications
| Vapor Pressure | 23hPa (at 20°C) |
| Analyte or Component Names | NO3 ion in H2O matrix |
| Density | 1.00035g/cm3 |
| Physical Form | Liquid |
| Molecular Formula | NO3 • H20 |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Solubility | Completely miscible |
| Element | NO3 ion |
| Solvent or Matrix | Water |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction