missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Nitrate Cuvette Tests

Pre-dosed reagents for maximum safety. Easy to use and photometric evaluation, packaged in a fully equipped box. Officially approved for consent limits. Environmentally friendly due to less chemicals.

Brand:  Hach Lange™ LCK340

Additional Details : Weight : 0.50000kg Transport : UN number : 3316 Chem class : 9 Pack group : II


 View more versions of this product

Product Code. 10012461

  • £145.00 / Pack of 25
Estimated Shipment: 19-08-2024
to see stock.

Description

Description

  • For use with DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments
  • ISO 7890-1-2-1986, DIN 38405 D9-2 certifications
  • Shelf life of 23 months from production date
  • Storage conditions of 15 - 25 °C
  • Measuring ranges of 0.23 - 13.5 mg/L NO3-N and 1.0 - 60.0 mg/L NO3
  • Precise and reliable measurement values
  • Convenient and error-free dosing of the reagents
Specifications

Specifications

25
15°C to 25°C
Nitrate Cuvette Test
5 to 35mg/L NO3-N
2.6-Dimethylphenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Substances and mixtures corrosive to metals Category 1
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers