Learn More
Mettler Toledo™ Nitrate Ion Selective Electrode
Brand: Mettler Toledo™ 51340800
Code : 59
Additional Details : Weight : 0.16000kg
Specifications
Polymer Membrane | |
0°C to 50°C | |
2 to 12 |