Learn More
Lovibond™ Nitrate/Nitrate-N Tube Test for SpectroDirect Spectrophotometer
Brand: Lovibond™ 2420702
Code : NEW
Additional Details : CAS Number : 7664-93-9 Weight : 0.21500kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Specifications
24 Tubes |
Safety and Handling
- nitrate-N tube test
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H290-May be corrosive to metals.
- H319-Causes serious eye irritation.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H318-Causes serious eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol, 2,6-xylenol, sulphuric acid, phosphoric acid