missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrate Standard for Ion Chromatography, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£40.00 - £107.00
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11410289
|
SPEX Certiprep
AS-NO39-2Y |
125 mL | Triple-leached LDPE bottle | N/A | |||||
10298772
|
SPEX Certiprep
AS-NO39-2Y |
125 mL | Triple-leached LDPE bottle |
£107.00
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11400289
|
SPEX Certiprep
AS-NO39-2X |
500 mL | Triple-leached LDPE bottle | N/A | |||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
NO3- | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7732-18-5 |
Specifications
7732-18-5 | |
0.1, 99.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
1000μg/mL | |
Nitrate Standard |
0.1, 99.9 | |
With Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
H2O |
Safety and Handling
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only