Learn More
Hanna Instruments™ Nitrate Test Reagents
High quality reagents that are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements.
Brand: Hanna Instruments™ HI93728-01
Description
- Reagents for the colorimetric determination of nitrate as nitrogen
- Pre-made reagents for ease of use
- Prepared with high purity chemicals
- High quality reagents that are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements
- Method is an adaptation of the cadmium reduction method in which the reaction between nitrate and the reagent causes an amber tint in the sample
Specifications
100 Pack |
Safety and Handling
- Nitrate Reagent
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 A
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P310-Immediately call a POISON CENTER/doctor/.
- P391-Collect spillage.
Supplemental information
- MIXTURE LIST-Contains : Potassium disulfate,Sulfanilic acid, Cadmium, powder, Copper sulfate pentahydrate