missing translation for 'onlineSavingsMsg'
Learn More
Nitric acid, 65-70%, 99.9999% (metals basis), Thermo Scientific Chemicals Product Code.: 11331578

Nitric acid, 65-70%, 99.9999% (metals basis), Thermo Scientific Chemicals

Product Code. 11331578
250 mL
Click to view available options
Quantity:
25 mL
250 mL
500 mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11331578

Brand: Thermo Scientific Alfa Aesar 010984.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Nitric acid, 65-70% is used in as a nitrating or an oxidizing agent in organic synthesis. It is industrially used for the manufacturing of fertilizers. Further, it finds use in manufacture of explosives, nylon precursors, specialty organic compounds and drug intermediates.It is also used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple. It is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxideShow MoreShow Less
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless to Yellow
Boiling Point 121°C
Concentration 65 to 70%
Density 1.41 g/mL
Odor Pungent
Quantity 250 mL
Physical Form Liquid
UN Number UN2031
Merck Index 14,6577
Solubility Information Miscible with water.
Formula Weight 63.01
Percent Purity 99.9999%
Assay (metals basis)
Chemical Name or Material Nitric acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number62500061 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog Number010984.AC,010984.AK,010984.AP
Lot Number62400561 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number010984.AC,010984.AK,010984.AP
Lot NumberS22J030 Certificate TypeCertificate of Analysis Date28/06/2023 Catalog Number010984.AC,010984,010984.AK,010984.AP
Lot NumberM05J017 Certificate TypeCertificate of Analysis Date18/01/2023 Catalog Number010980.04,010984.AK,010980.18,010980.09
Lot NumberS22I035 Certificate TypeCertificate of Analysis Date23/11/2022 Catalog Number010984.AC,010984.AK,010989.AU,010984.AP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nitric acid, 65-70%, 99.9999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.