missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid 68-70% d=1.42, Certified AR, for Analysis, Fisher Chemical™

Product Code. 10378433
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

HNO3, CAS Number-7697-37-2, acide nitrique, azotic acid, hydrogen nitrate, salpetersaeure, rfna, nitryl hydroxide, nitrous fumes, nital, acidum nitricum, aqua fortis, 2.5L, 122 deg.C, CHEBI:48107, Colorless Yellow, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -42 deg.C, 11349, 63.012

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH 1
Color Yellow
Melting Point -42°C
Boiling Point 122°C
Packaging Plastic Coated Glass bottle
Quantity 2.5 L
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Nitric acid SG 1.42 (70%)
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.