missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid 68% d=1.42, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™

Product Code. 10149982
Click to view available options
Quantity:
500 mL
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

HNO3, CAS Number-7697-37-2, hydrogen nitrate, nital, aqua fortis, azotic acid, acide nitrique, acidum nitricum, rfna, salpetersaeure, nitrous fumes, nitryl hydroxide, 2.5L, CHEBI:48107, Yellow, 63.02g/mol, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -41 deg.C, 11349, 63.012, HDPE plastic bottle

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH 1
Color Yellow
Melting Point -41°C
Packaging HDPE plastic bottle
Vapor Pressure 0.94kPa (20°C)
Quantity 1 L
Physical Form Liquid
Formula Weight 63.02g/mol
Chemical Name or Material Nitric Acid
compliance-icons compliance-icons
Product Identifier
  • Nitric acid (>= 68%)
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.