Learn More
Nitric acid, 70%, Thermo Scientific Chemicals
May darken on storage
£31.36 - £65.05
Chemical Identifiers
CAS | 7697-37-2, 7732-18-5 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15533560
|
Thermo Scientific Acros
124650010 |
1 L |
£31.36
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15528251
|
Thermo Scientific Acros
124650025 |
2.5 L |
£65.05
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7697-37-2, 7732-18-5 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
O[N+]([O-])=O |
Specifications
7697-37-2, 7732-18-5 | |
122.0°C | |
1.4 | |
HNO3 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01 | |
Pure |
-42.0°C | |
1.4000g/mL | |
HNO3 | |
MFCD00011349 | |
Solubility in water: >100 g/100mL (20°C). | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
70% | |
Nitric acid |
Safety and Handling
GHS H Statement May intensify fire; oxidizer. Causes severe skin burns and eye damage. May be corrosive to metals. Corrosive to the respiratory tract.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Keep/Store away from clothing/ combustible materials.
Danger
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.