missing translation for 'onlineSavingsMsg'
Learn More
Nitric acid, ≥99% (T), Honeywell™ Product Code.: 15627300
compliance-icons compliance-icons compliance-icons

Nitric acid, ≥99% (T), Honeywell™

Product Code. 15627300
500 mL, Glass bottle
Click to view available options
Quantity:
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 15627300

Brand: Honeywell Research Chemicals 84392500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

purum p.a., fuming, packed in coated, shock- and leak-protected glass bottle,≥99% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Specifications

pH <1 (at 20°C)
Color Colorless
CAS 7697-37-2
Melting Point -41.59°C
Boiling Point 120.5°C
Density 1.48g/cu.cm at 20°C
Packaging Glass bottle
Vapor Pressure 8mmHg (20°C)
Quantity 500 mL
Molecular Formula HNO3
Physical Form Liquid
MDL Number MFCD00011349
UN Number UN2031A
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
Solubility Information 1000g/L - completely soluble
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Formula Weight 63.01g/mol
Percent Purity ≥99% (T)
Chemical Name or Material Nitric acid, purum p.a., fuming, packed in coated, shock- and leak-protected glass bottle
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nitric acid
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nitric acid, ≥99% (T), Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.