missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid, ACS, 68.0-70.0%, Thermo Scientific Chemicals

Product Code. 15431327
Click to view available options
Quantity:
3178 g
6 x 3178 g
Unit Size:
1kg
Each
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 15431327

Brand: Thermo Scientific Alfa Aesar 033260.X#

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Main commercial usage of Nitric acid, ACS, 68.0-70.0% is in the production of fertilizers and reagent for nitration. It is used in the applications for the production of explosives, nylon precursors and specialty organic compounds. Nitric acid is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions. It is used as a chemical doping agent for organic semiconductors and in purification processes for raw carbon nanotubes, artificially age pine and maple.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Appearance Colorless to pale yellow liquid
Color Colorless to Yellow
Boiling Point 121°C
Density 1.41 g/cm3 (at 20°C)
Odor Pungent
Vapor Pressure 23 hPa (17 mm Hg) (at 20°C)
Assay Percent Range 68.0 to 70.0%
Quantity 6 x 3178 g
Physical Form Liquid
Synonym hydrogen nitrate, aqua fortis, azotic acid
Solubility Information Miscible with water and ether.
Formula Weight 63.01
Grade ACS
Chemical Name or Material Nitric acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.