missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Specifications
Color | Colorless |
Melting Point | -42.0°C |
Boiling Point | 122.0°C |
CAS Min % | 64.0, 34.0 |
CAS Max % | 66.0, 36.0 |
Density | 1.4000g/mL |
Packaging | Metal Drum |
Specific Gravity | 1.4 |
Quantity | 25 L |
Linear Formula | HNO3 |
Show More |
Safety and Handling
Product Identifier
- Nitric acid, ca. 65% solution in water
- Danger
- Oxidising liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- EUH071-Corrosive to the respiratory tract.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction