missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, DILUT-IT Concentrate, 1.0M, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
For dilution to 1 L
Packaging:
LDPE ampule
Unit Size:
Each
Description
DILUT-IT solutions are relied upon for fast, accurate results in life sciences and pharmaceutical quality control applications. Simply dilute the volumetric concentrate to 1 liter (carefully following instructions provided with the package) and the resulting solution will have an exact normality within ±0.1%.
- Enhanced productivity: Eliminate expensive, time-consuming steps, such as the selection and analysis of starting reagents, and weighing and standardization of the final solution
- Space saving: Products require only one-tenth of the space required for ready-made solutions
- Reduced shipping costs: Spend less on shipping due to the reduced weight and package size of a concentrate
Specifications
Specifications
| Color | Colorless |
| CAS | 7697-37-2 |
| Melting Point | 0°C |
| Boiling Point | 83°C |
| Concentration | 1mol/L |
| Density | 1.2g/cm3 |
| Odor | Pungent |
| Packaging | LDPE ampule |
| Quantity | For dilution to 1 L |
| Molecular Formula | HNO3 |
| Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction