Learn More
Nitric acid, Environmental Grade Plus, 68-70%, Thermo Scientific™
£449.00 - £924.00
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
O[N+]([O-])=O |
Specifications
Colorless to Yellow | |
121°C | |
Pungent | |
Liquid, Double Sub-Boiling Quartz Distillation | |
UN2031 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
68 to 70% | |
Nitric acid |
7697-37-2 | |
1.41 g/mL | |
HNO3 | |
MFCD00011349 | |
14,6577 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01 | |
Environmental Plus |
Safety and Handling
GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.
P210-P220-P221-P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
H272-H290-H314-H331
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: NITRIC ACID
missing translation for 'einecsNumber' : 231-714-2
missing translation for 'rtecsNumber' : QU5775000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.