Learn More
Nitric acid, extra pure, 60% solution in water, Thermo Scientific Chemicals
£32.86 - £65.33
Chemical Identifiers
CAS | 7697-37-2, 7732-18-5 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10306690
|
Thermo Scientific Acros
133620010 |
1 L | Glass Bottle |
£32.86
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10027192
|
Thermo Scientific Acros
133620011 |
1 L | Plastic bottle |
£33.77
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10295230
|
Thermo Scientific Acros
133620025 |
2.5 L | Glass Bottle |
£64.72
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10631575
|
Thermo Scientific Acros
133620026 |
2.5 L | Plastic bottle |
£65.33
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7697-37-2, 7732-18-5 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
O[N+]([O-])=O |
Specifications
Colorless to Yellow | |
-42.0°C | |
59.0, 37.0 | |
1.3300g/mL | |
HNO3 | |
Liquid | |
01,733; 03,212; 04,356; 05,474 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01 | |
Extra Pure |
7697-37-2, 7732-18-5 | |
122.0°C | |
63.0, 41.0 | |
1.33 | |
HNO3 | |
MFCD00011349 | |
15, 6663 | |
Solubility in water: >100 g/100mL (20°C). | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
59.0 to 63.0% | |
Nitric acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage. May be corrosive to metals. Corrosive to the respiratory tract.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Danger
- Nitric acid, 20-64%
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only