missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, puriss., Honeywell Fluka™
Puriss., 64-66%
£32.41 - £76.28
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15600840
|
Honeywell-Fluka
07006-1L |
1 L |
£32.41
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15610840
|
Honeywell-Fluka
07006-2.5L |
2.5 L |
£76.28
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxideShow More | |
CHEBI:48107 | |
O[N+]([O-])=O |
Specifications
7697-37-2 | |
1.37 to 1.41g/mL at 20°C | |
8mmHg (20°C) | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
puriss. |
64 to 66% | |
≤0.0005% (as SO4) | |
HNO3 | |
2031 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01g/mol | |
Nitric Acid |
Safety and Handling
P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
H272-H290-H314
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Nitric acid
Signal Word
- Danger
Hazard Category
- Oxidising liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : nitric acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nitric Acid, puriss., Honeywell Fluka™