missing translation for 'onlineSavingsMsg'
Learn More
Nitric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone, ≥65%, Honeywell Fluka™ Product Code.: 15643090
compliance-icons compliance-icons compliance-icons

Nitric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone, ≥65%, Honeywell Fluka™

Product Code. 15643090 Shop All Honeywell Fluka Products
2.5 L, Plastic Bottle
Click to view available options
Quantity:
1 L
2.5 L
40 kg
4 x 2.5 L
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Poly Bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 15643090

Brand: Honeywell-Fluka 307092.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone,≥65%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxideShow MoreShow Less
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Density 1.37 to 1.41g/mL at 20°C
Packaging Plastic Bottle
Residue after Ignition ≤0.0005% (as SO4)
Vapor Pressure 8mmHg (20°C)
Quantity 2.5 L
UN Number 2031
Formula Weight 63.01g/mol
Percent Purity ≥65%
Grade puriss. p.a.
Chemical Name or Material Nitric Acid, Reag. Ph. Eur. reag. ISO
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nitric acid
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.,Do not breathe dust/fume/gas/mist/vapours/spray.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.
  • P234-Keep only in original container.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.,IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.,IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.,IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.,IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.,IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains :
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nitric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone, ≥65%, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.