missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid solution 8mol/L, Fisher Chemical™
missing translation for 'orderingAttributeHoverText'
Quantity:
1000 mL
Packaging:
Bottle
Specifications
Specifications
CAS | 7697-37-2 |
Concentration | 8M (8N) |
Packaging | Bottle |
Quantity | 1000 mL |
Molecular Formula | HNO3 |
MDL Number | MFCD00011349 |
Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
SMILES | O[N+]([O-])=O |
IUPAC Name | nitric acid |
Show More |
Safety and Handling
Product Identifier
- Nitric acid 8M Volumetric Solution
- Danger
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'