missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid Solution, Honeywell™ Fluka™
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Colorless |
CAS | 7697-37-2 |
Boiling Point | 100°C |
Odor | Pungent |
Packaging | Poly bottle |
Quantity | 1 L |
Molecular Formula | HNO3 |
Physical Form | Clear liquid |
MDL Number | MFCD00011349 |
UN Number | UN2031 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'