missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrilotriacetic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- Nitrilotriacetic acid (2,2',2-nitrilotriacetic acid) is a tricarboxylic acid used for chemical purification
Applications
- A known mutagen, suitable for cell culture experiments of DNA damage
- Nitrilotriacetic acid can be used for protein elution using the His-tag method and chemical chelation
- Nitrilotriacetic acid is capable of chelating Ca2+, Co2+, Cu2+, and Fe3+
- Intended for sensitive applications such as HPLC
Specifications
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 245.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Plastic bottle |
Linear Formula | N(CH2CO2H)3 |
Quantity | 250 g |
Beilstein | 04,369 |
Show More |
Safety and Handling
Product Identifier
- Nitrilotriacetic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'