missing translation for 'onlineSavingsMsg'
Learn More

Nitrite Standard Solution, Chem Lab™

Nitrite standard solution 1000 µg/ml for IC (Ion HIQU)

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O
View More Specs

Description

Description

Specifications

Chemical Identifiers

7632-00-0
69.00
sodium nitrite
NNaO2
LPXPTNMVRIOKMN-UHFFFAOYSA-M
[Na+].[O-]N=O

Specifications

Solution contains 1000 µg/ml of NO2-
Colorless
NNaO2
[Na+].[O-]N=O
69.00
100 C
Liquid
7632-00-0
Reference standards acc. NIST SRM 136f
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium nitrite
Odorless
Soluble in water
Videos
SDS
Documents

Documents

Product Certifications