Learn More
Ansell™ BioClean™ Clearview™ Autoclavable Goggles
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
390.98 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Goggles
Description
- Non-Sterile and Sealed in a PE Bag to maintain cleanliness
- Upper & Indirect Ventilation holes
- EN 166:2001 Optical Class 1
- Toughened Anti-Scratch Polycarbonate lens
- Tested to over 40 Autoclave Cycles
- Super-soft thermoplastic provides unrivaled comfort
- Processed and packed in our NEBB Certified ISO Class 4 (Class 10) Cleanroom
- Replacement lens available (BCAG-SL 19-668-191)
Specifications
Certifications/Compliance | ISO 9001:2015, (EU)2016/425, EN 166:2001 |
Lens Coating | Scratch-resistant |
Product Type | Autoclavable Goggles |
No. per Case | 12/Cs. |
No. per Pack | 1 Ea. |
Ventilation Details | Indirect base ventilation |
Material | Polycarbonate |