missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean™ MTA™ Sterile Cleanroom Tie-On Face Mask MTA 210-1
BIOPHARMA

Low-linting sterile face mask provides high particle and bacterial filtration efficiency while providing comfort and breathability.

Brand:  Ansell MTA210-1

450.76 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


Product Code. 12775925

  • £697.00 / Pack of 500
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Low-linting fabric
  • ISO Class 4 compatible
  • High bacterial and particle filtration efficiency
  • 210 mm, universal size
  • Fully enclosed, malleable steel nose-band
  • Non-woven polypropylene ties
Applications: Cleanroom, Pharma, Controlled Environments, PPE, 503B Facilities/Compounding, Manufacturing, Industrial
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cleanroom Face Mask
Ties
Standard
Pleated with Ties
Class 10 (ISO 4)
>97% (PFE), >96.5% (BFE), 97.9% (VFE)
8.3 in.
MTA210-1
Individually Bagged
White
Polypropylene
Sterile
ASTM F2101, ISO 2859
Single-use
Yes
21 cm
One piece per sealed inner PE bag; 50 inner bags per sealed outer PE bag; 10 outer bags per lined carton (500 pieces)
BioClean™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers