Learn More
Nitrobenzene, 99%, Extra Pure, Thermo Scientific Chemicals
£41.10
Chemical Identifiers
CAS | 98-95-3 |
---|---|
Molecular Formula | C6H5NO2 |
Molecular Weight (g/mol) | 123.11 |
MDL Number | MFCD00007043 |
InChI Key | LQNUZADURLCDLV-UHFFFAOYSA-N |
Synonym | nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene |
PubChem CID | 7416 |
ChEBI | CHEBI:27798 |
IUPAC Name | nitrobenzene |
SMILES | C1=CC=C(C=C1)[N+](=O)[O-] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
98-95-3 | |
123.11 | |
LQNUZADURLCDLV-UHFFFAOYSA-N | |
7416 | |
nitrobenzene |
C6H5NO2 | |
MFCD00007043 | |
nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene | |
CHEBI:27798 | |
C1=CC=C(C=C1)[N+](=O)[O-] |
Specifications
98-95-3 | |
100.0 | |
Yellow | |
210.0°C to 211.0°C | |
Authentic | |
C6H5NO2 | |
1.5503 to 1.5523 | |
05, 233 | |
15, 6674 | |
nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene | |
LQNUZADURLCDLV-UHFFFAOYSA-N | |
nitrobenzene | |
7416 | |
123.11 | |
Extra Pure | |
Nitrobenzene |
98.5 | |
5.0°C to 6.0°C | |
1.2050g/mL | |
88°C | |
98.5% min. (GC) | |
C6H5NO2 | |
MFCD00007043 | |
02,295; 07,251; 08,358; 17,16 | |
1.205 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,benzene,ether and oils | |
C1=CC=C(C=C1)[N+](=O)[O-] | |
123.11 | |
CHEBI:27798 | |
99% | |
Liquid |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Toxic in contact with skin.
Harmful to aquatic life with long lasting effects.
Toxic if sw
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-716-
- Nitrobenzene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H351-Suspected of causing cancer.
- H360F-May damage fertility.
- H372-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : nitrobenzene
RUO – Research Use Only