missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, 99%, Reagent Grade, Honeywell™

Reagent Grade, 99%

£44.49 - £990.29

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.111
MDL Number MFCD00007043
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
PubChem CID 7416
ChEBI CHEBI:27798
IUPAC Name nitrobenzene
SMILES C1=CC=C(C=C1)[N+](=O)[O-]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15637980
View Documents
Honeywell Research Chemicals
N10950-1L
1 L Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15647980
View Documents
Honeywell Research Chemicals
N10950-2.5L
2.5 L Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15627980
View Documents
Honeywell Research Chemicals
N10950-10L
10 L Stainless steel can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

98-95-3
123.111
LQNUZADURLCDLV-UHFFFAOYSA-N
7416
nitrobenzene
C6H5NO2
MFCD00007043
nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
CHEBI:27798
C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

98-95-3
Yellow
8.0 to 8.5 (at 1.0000g/L 20.0°C)
0.4
899°F
C6H5NO2
UN1662
nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
C1=CC=C(C=C1)[N+](=O)[O-]
123.111
4.2 (vs air)
123.11g/mol
0.99
Nitrobenzene
5°C to 6°C
1.196g/mL at 25°C
210°C to 211°C
88°C (190.4°F)
Pungent
MFCD00007043
507540
LQNUZADURLCDLV-UHFFFAOYSA-N
nitrobenzene
7416
CHEBI:27798
0.15mmHg (20°C), 50mmHg (120°C)
Liquid
Videos
Safety and Handling

Safety and Handling

Missing Phrase - N15.00950417-P201-P280-P302 + P352 + P312-P304 + P340 + P312-P308 + P313

missing translation for 'dotInformation' : 6.1/PGII

missing translation for 'rtecsNumber' : 202-716-0

SDS
Documents

Documents

Product Certifications