missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene-d5, for NMR, 99% atom D, Thermo Scientific Chemicals

Product Code. 10144180
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4165-60-0
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 128.14
MDL Number MFCD00044415
InChI Key LQNUZADURLCDLV-RALIUCGRSA-N
Synonym nitrobenzene-d5, benzene-d5, nitro, nitrobenzene d5, benzene-d5-, nitro, 1-nitro 2 h? benzene, nitrobenzene-d5, 99.5 atom % d, nitro 2h5 benzene, 1-nitro 2h5 benzene, 2h5 benzene, nitro, nitrobenzene-d5, analytical standard
PubChem CID 123210
IUPAC Name 1,2,3,4,5-pentadeuterio-6-nitrobenzene
SMILES [2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])[N+]([O-])=O

Specifications

Physical Form Liquid
Color Colorless to Yellow
Deuterium (D) 98.5% min.
Packaging Glass Bottle
Quantity 10 g
Linear Formula C6D5NO2
Formula Weight 128.15
Grade NMR
Chemical Name or Material Nitrobenzene-d5, 99 atom % D
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nitrobenzene-d5
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H351-Suspected of causing cancer.
  • H361f-Suspected of damaging fertility.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'