missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva™ Whatman™ NC45 Nitrocellulose Filtration Membranes
Whatman™ NC45 Nitrocellulose Membrane Filters are known for higher strength and flexibility, low extractables, and narrow pore size distribution. They are recommended for routine applications and are manufactured under strictly controlled conditions.
Brand: Cytiva™ 10401122
52.81 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Product Type: Filtration Membrane
- Grade: NC45
- Material: Nitrocellulose
- Diameter: 85 mm
- Pore Size: 0.45 μm
- For Use With (Application): Microbiological Studies, Sample Preparation
- Quantity: 50 Pk.
Specifications
Autoclavable | |
Nitrocellulose | |
0.45 μm | |
Microbiological Studies, Sample Preparation | |
NC45 | |
50 Pk. | |
Membrane Filter | |
85 mm |
Reusable | |
Nitrocellulose | |
Non-sterile | |
NC45 | |
50Pack | |
Circular | |
3.3 in. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- NC45 membrane, cellulose nitrate, 0.45µm 85mm
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose