Learn More
Thermo Scientific Chemicals Nitrocellulose stripping buffer (10X)
Brand:  Thermo Scientific Chemicals J62541.AK
 View more versions of this product
Product Code. 15414909
  • £50.45 / 250mL
Explore more special offers
Description
Description
Ideal for removal of secondary antibody after detection of highly abundant targets. Nitrocellulose Stripping Buffer may perform well with nitrocellulose membranes or antibody-dye conjugates. It is also employed for nucleic acid and protein related blotting works.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot Stripping Buffer
Liquid
250 mL
10X