Learn More
Lenz Laborglasintrumente Nitrogen Tubes (Schlenk Tubes) with NS Stopcock
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
£86.85 - £97.95
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | ||||||
10613442
|
Lenz Laborglasintrumente
03370149 |
NS 14/23 Socket |
£86.85
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||||
10376211
|
Lenz Laborglasintrumente
03370249 |
NS 14/23 Cone |
£97.95
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
Description
- With conical ground joint and NS stopcock
- Made of DURAN™ tubing
Specifications
250 mL | |
Duran Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasintrumente Nitrogen Tubes (Schlenk Tubes) with NS Stopcock