missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitromethane, 99+%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Melting Point | -29.0°C |
Color | Colorless to Yellow |
Density | 1.1200g/mL |
Boiling Point | 100.0°C to 102.0°C |
Flash Point | 35°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Nitromethane
- Warning
- Acute toxicity Category 4
- Flammable liquid Category 3
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'