missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitronium hexafluorophosphate, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Beschreibung
Nitronium hexafluorophosphate is a nitrating reagent used in the nitration of alkanes such as methane, ethane, propane, n-butane, isobutane, neopentane and cyclohexane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spezifikation
Color | White |
Physical Form | Crystalline |
Quantity | 5 g |
Solubility Information | Soluble in dichloromethane and nitroethane. |
Formula Weight | 190.97 |
Sensitivity | Moisture sensitive |
Appearance | Off-white Cristals |
Chemical Name or Material | Nitronium hexafluorophosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'