Learn More
Nitroso-R-Salt, Extra Pure, SLR, Determination Of Cobalt, Fisher Chemical
Brand: Fisher Chemical N/3650/46
Code : 27
Additional Details : CAS Number : 525-05-3 Weight : 0.02500kg
Description
25g, Plastic powder jar
Specifications
525-05-3 | |
Orange to Yellow | |
C10H5NNa2O8S2 | |
25 g | |
DMKMTGULLYISBH-UHFFFAOYSA-L | |
disodium;3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate | |
10672 | |
Powder |
300°C | |
Plastic powder jar | |
MFCD00149246 | |
nitroso-r salt, nitroso r, sodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate, disodium 1-nitroso-2-naphthol-3,6-disulfonate, nitroso r salt, 1-nitroso-3,6-disulfonate, 2,7-naphthalenedisulfonic acid, 3-hydroxy-4-nitroso-, disodium salt, 3-hydroxy-4-nitroso-2,7-naphthalenedisulfonic acid disodium salt, 1-nitroso-2-naphthol-3,6-disulfonic acid disodium salt, disodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulphonate | |
[Na+].[Na+].OC1=C(N=O)C2=CC=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O | |
377.25 | |
377.25g/mol |
Safety and Handling
- 1-Nitroso-2-naphthol-3,6-disulfonic acid, disodium salt hydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.