missing translation for 'onlineSavingsMsg'
Learn More

Nitrosonium hexafluorophosphate, 95%, Thermo Scientific Chemicals

£78.69 - £154.77

Chemical Identifiers

CAS 16921-91-8
Molecular Formula F6NOP
Molecular Weight (g/mol) 174.97
MDL Number MFCD00040324
InChI Key SAKPNYRUBHJGAR-UHFFFAOYSA-N
Synonym nitrosonium hexafluorophosphate, nitrilooxonium hexafluorophosphate, nopf6, nitrosyl hexafluorophosphate, nitrosonium hexafluorophosphat, nitrosylhexafluorophosphate, azanylidyneoxidanium hexafluorophosphate, hexafluoro-$l^ 5-phosphanuide; nitrosonium
PubChem CID 10910083
IUPAC Name azanylidyneoxidanium;hexafluorophosphate
SMILES N#[O+].F[P-](F)(F)(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10655073
SDS View Documents
Thermo Scientific Acros
383730050
5 g Plastic bottle
£78.69
5g
Please to purchase this item. Need a web account? Register with us today!
10556013
SDS View Documents
Thermo Scientific Acros
383730250
25 g Plastic bottle
£154.77
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nitrosonium hexafluorophosphate, 95%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.