missing translation for 'onlineSavingsMsg'
Learn More

Nitrosonium hexafluorophosphate, 96%

Product Code. 11312248
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11312248

Brand: Thermo Scientific Alfa Aesar 013149.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16921-91-8 | F6NOP | 174.97 g/mol

Nitrosonium hexafluorophosphate is used as initiator in the polymerization reactions of 1,2-epoxides such as styrene oxide, cyclohexene oxide and 1,2-epoxybutane. Further, it is involved in the polymerization of styrene, n-butyl vinyl ether and tetrahydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16921-91-8
Molecular Formula F6NOP
Molecular Weight (g/mol) 174.97
MDL Number MFCD00040324
InChI Key SAKPNYRUBHJGAR-UHFFFAOYSA-N
Synonym nitrosonium hexafluorophosphate, nitrilooxonium hexafluorophosphate, nopf6, nitrosyl hexafluorophosphate, nitrosonium hexafluorophosphat, nitrosylhexafluorophosphate, azanylidyneoxidanium hexafluorophosphate, hexafluoro-$l^ 5-phosphanuide; nitrosonium
PubChem CID 10910083
IUPAC Name azanylidyneoxidanium;hexafluorophosphate
SMILES N#[O+].F[P-](F)(F)(F)(F)F

Specifications

Physical Form Crystalline
Quantity 25 g
Linear Formula NOPF6
UN Number UN3260
Solubility Information Soluble in acetonitrile.
Formula Weight 174.96
Percent Purity 96%
Odor Odorless
Sensitivity Moisture sensitive
Chemical Name or Material Nitrosonium hexafluorophosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.