missing translation for 'onlineSavingsMsg'
Learn More

Nitrosonium tetrafluoroborate, 98%

Codice prodotto. 11472468
missing translation for 'orderingAttributeHoverText'
Quantity:
2 g
10 g
50 g
missing translation for 'unitSize'
10g
2g
50g
Questo articolo non è restituibile. Consulta la politica di reso

Codice prodotto. 11472468

missing translation for 'mfr': Thermo Scientific Alfa Aesar A15806.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Questo articolo non è restituibile. Consulta la politica di reso

CAS: 14635-75-7 | BF4HNO2 | 133.82 g/mol

Nitrosonium tetrafluoroborate is a nitrosating agent and oxidizer used in organic synthesis. It is also used to convert arylamines directly to diazonium tetrafluoroborates, which undergo the Balz-Schiemann reaction in situ to give high yields of aryl fluorides. Further, it is used as a catalyst for iodination of aromatics by iodide ion with air as oxidant, conversion of secondary amines to nitrosamines and in deamination of primary amides. It is involved in the conversion of arylhydrazines to azides. In addition to this, it reacts with ferrocene to prepare ferrocenium tetrafluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nitrosonium tetrafluoroborate is a nitrosating agent and oxidizer used in organic synthesis. It is also used to convert arylamines directly to diazonium tetrafluoroborates, which undergo the Balz-Schiemann reaction in situ to give high yields of aryl fluorides. Further, it is used as a catalyst for iodination of aromatics by iodide ion with air as oxidant, conversion of secondary amines to nitrosamines and in deamination of primary amides. It is involved in the conversion of arylhydrazines to azides. In addition to this, it reacts with ferrocene to prepare ferrocenium tetrafluoroborate.

Solubility
Soluble in acetonitrile.

Notes
Hygroscopic. Air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids and amines.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 14635-75-7
Molecular Formula BF4HNO2
Molecular Weight (g/mol) 133.82
MDL Number MFCD00011433
InChI Key XYRQURYIJKJVSF-UHFFFAOYSA-N
Synonym nitrosonium tetrafluoroborate, nitrosyl tetrafluoroborate, nitrosyltetrafluoroborate, nobf4, nitrilooxonium tetrafluoroborate, azanylidyneoxidanium tetrafluoroborate
PubChem CID 11137142
IUPAC Name azanylidyneoxidanium;tetrafluoroborate
SMILES [O-][NH+]=O.F[B-](F)(F)F

Specifica

Boiling Point 250°C (0.01 mmHg)(Sublimation)
Melting Point 250°C (Sublimation)
Quantity 10 g
UN Number UN3260
Merck Index 14,6649
Solubility Information Soluble in acetonitrile.
Formula Weight 116.82
Percent Purity 98%
Odor Irritating
Sensitivity Moisture sensitive
Density 2.185
Chemical Name or Material Nitrosonium tetrafluoroborate
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Nitrosonium tetrafluoroborate
Avvertenza
  • Danger
Categoria di pericolo
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Indicazioni di pericolo
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informazioni supplementari
  • MIXTURE LIST-Contains : Nitrosonium tetrafluoroborate

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato