missing translation for 'onlineSavingsMsg'
Learn More

NKH 477, Tocris Bioscience™

Product Code. 30001270
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Water-soluble adenylyl cyclase activator

Water-soluble analog of forskolin (Cat. No. 1099) that is a potent activator of adenylyl cyclase; shows some selectivity for cardiac (type V) adenylyl cyclase. Stimulates bronchodilation (EC50 = 32.6 nM) and is an orally active potent positive chronotrope and hypotensive agent in vivo.

Chemical Identifiers

CAS 138605-00-2
Molecular Formula C27H43NO8
Molecular Weight (g/mol) 509.64
InChI Key RSOZZQTUMVBTMR-XGUNBQNXSA-N
Synonym colforsin daropate, colforsin dapropate hcl, colforsin dapropate, colforsin daropate hcl, beta-alanine, n,n-dimethyl-, 3r,4ar,5s,6s,6as,10s,10ar,10bs-5-acetyloxy-3-ethenyldodecahydro-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-1h-naphtho 2,1-b pyran-6-yl ester, 3r,4ar,5s,6s,6as,10s,10ar,10bs-5-acetyloxy-3-ethenyl-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2h-benzo f chromen-6-yl 3-dimethylamino propanoate
PubChem CID 444029
IUPAC Name [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-5-acetyloxy-3-ethenyl-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-6-yl] 3-(dimethylamino)propanoate
SMILES CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)OC(=O)CCN(C)C

Specifications

Formula Weight 546.1
Percent Purity >98%
Quantity 10 mg
Chemical Name or Material NKH 477
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.