missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKTR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567992
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Human samples.
This gene encodes a membrane-anchored protein with a hydrophobic amino terminal domain and a cyclophilin-like PPIase domain. It is present on the surface of natural killer cells and facilitates their binding to targets. Its expression is regulated by IL2 activation of the cells.Specifications
NKTR | |
Polyclonal | |
Unconjugated | |
Nktr | |
5330401F18Rik; cyclophilin-related protein; D9Wsu172e; natural killer cell triggering receptor; natural killer triggering receptor; natural killer tumor recognition protein; natural killer tumor recognition sequence; natural killer tumor recognition sequence-like; natural killer-tumor recognition sequence; natural-killer cells cyclophilin-related protein; NKTR; NK-TR protein; NK-tumor recognition protein; p104; Peptidyl-prolyl cis-trans isomerase NKTR; PPIase; Rotamase | |
Rabbit | |
affinity chromatography | |
RUO | |
4820 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.02% sodium azide; pH 7.3 | |
P30414 | |
Nktr | |
Synthesized peptide derived from human NKTR. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |