missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2.2 Mouse, Clone: NX2/1524, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4821-MSM4-P1
This item is not returnable.
View return policy
Specifications
NKX2.2 | |
Nuclear | |
NX2/1524 | |
Unconjugated | |
XP_006723629 | |
Mouse | |
40-50kDa | |
Protein A/G Purified | |
RUO | |
4821 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
NKX2-2 | |
Human recombinant NKX2.2 protein fragment (aa1-119) | |
Pancreas or Ewing s sarcoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |