missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nkx3.1 Monoclonal Antibody (0361), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA116906
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: antigen standard for NKX3 (transient overexpression lysate), LNCAP cell extracts, mouse testis protein, mouse testis tissue lysate.
The homeodomain-containing transcription factor NKX3-1 is a putative prostate tumor suppressor that is expressed in a largely prostate-specific and androgen-regulated manner. Loss of NKX3-1 protein expression is a common finding in human prostate carcinomas and prostatic intraepithelial neoplasia.Specifications
Nkx3.1 | |
Monoclonal | |
2 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P97436, Q99801 | |
NKX3-1 | |
Antibody has been raised against human Nkx3.1 full-length protein produced in bacteria as an hexahistidine-tagged protein. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0361 | |
Unconjugated | |
NKX3-1 | |
bagpipe; BAPX2; Bax; Drosophila NK-3 transcription factor, locus 1; homeobox protein NK-3 homolog A; Homeobox protein Nkx-3.1; NK homeobox, family 3, A; NK3 homeobox 1; NK3 transcription factor homolog A; NK3 transcription factor related, locus 1; NK-3 transcription factor, locus 1; NK-3 transcription factor, locus 1 (Drosophila); NKX3; NKX3.1; Nkx-3.1; Nkx3-1; NKX3-1 antibody; NKX3A; prostate-specific ag | |
Mouse | |
Protein G | |
RUO | |
18095, 305999, 4824 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |