Learn More
NM107, MedChemExpress
MedChemExpress NM107 (2'-C-Methylcytidine) is an nucleoside inhibitor of the hepatitis C virus (HCV) NS5B polymerase, the EC50 of NM107 in the wild-type replicon cells is 1.85 μM.
Brand:  Medchem Express HY-10468
Encompass_Preferred
 View more versions of this product
Product Code. 16416707
  • £82.28 / 5mg
Explore more special offers
Description
Description
NM107 (2'-C-Methylcytidine) is an nucleoside inhibitor of the hepatitis C virus (HCV) NS5B polymerase, the EC50 of NM107 in the wild-type replicon cells is 1.85 μM.
Specifications
Specifications
NM107
White
C10H15N3O5
2'-C-Methylcytidine NM-107
NC(C=CN1[C@@H]2O[C@H](CO)[C@@H](O)[C@@]2(C)O)=NC1=O
257.24
Research
COVID-19-anti-virus
20724-73-6
Research
5 mg
DMSO : 100 mg/mL (388.74 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (194.37 mM)
257.24
98.9%
Solid