missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NNC 26-9100, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
NNC 26-9100 is a somatostatin sst4 receptor agonist that displays > 100-fold selectivity over sst2 receptors (Ki values are 6 and 621 nM for sst4 and sst2 receptors respectively). Potently inhibits forskolin -induced cAMP accumulation (EC50 = 26 nM).
Specifications
Specifications
CAS | 199522-35-5 |
Molecular Formula | C22H25BrCl2N6S |
Quantity | 10 mg |
Synonym | 1-3-n-5-bromopyridin-2-yl-n-3,4-dichlorobenzyl amino propyl-3-3-1h-imidazol-4-yl propyl thiourea, 1-3-5-bromo-pyridin-2-yl-3,4-dichloro-benzyl-amino-propyl-3-3-1h-imidazol-4-yl-propyl-thiourea, 1-3-5-bromopyridin-2-yl 3,4-dichlorophenyl methyl amino propyl-3-3-1h-imidazol-4-yl propyl thiourea, thiourea,n-3-5-bromo-2-pyridinyl 3,4-dichlorophenyl methyl amino propyl-n'-3-1h-imidazol-5-yl propyl |
InChI Key | UREJDUPKGMFJRU-UHFFFAOYSA-N |
SMILES | C1=CC(=C(C=C1CN(CCCNC(=S)NCCCC2=CN=CN2)C3=NC=C(C=C3)Br)Cl)Cl |
IUPAC Name | 1-[3-[(5-bromopyridin-2-yl)-[(3,4-dichlorophenyl)methyl]amino]propyl]-3-[3-(1H-imidazol-5-yl)propyl]thiourea |
Molecular Weight (g/mol) | 556.348 |
PubChem CID | 9893924 |
Formula Weight | 556.35 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'