missing translation for 'onlineSavingsMsg'
Learn More

NNMT Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA5121872

Product Code. 17703274

  • £140.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver. This gene encodes the protein responsible for this enzymatic activity which uses S-adenosyl methionine as the methyl donor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NNMT
Polyclonal
Unconjugated
NNMT
Nicotinamide N-methyltransferase; NNMT
Rabbit
Antigen affinity chromatography
RUO
4837
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300
P40261
NNMT
Recombinant protein Nicotinamide-N-Methyltransferase. The antigen corresponds to amino acid range 1-258 of the target protein.
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.