missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan No admittance to unauthorised persons sign, aluminum, 100mm x 300mm
Brand: Manutan A346024
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- No admittance to unauthorised persons
- aluminum
- Dimensions (H x W) : 100mm x 300mm