missing translation for 'onlineSavingsMsg'
Learn More

Manutan No entry asbestos barrier tape, 100m x 150mm

Brand:  Manutan A353865

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12825898

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • No entry asbestos
  • Dimensions (L x W) : 100m x 150mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers